Läs mer
Triethyl 3-Methyl-4-phosphono-2-butenoate (~90%), TRC
Produktkod. 30451922
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30451922 5 g
1 options
Produktkod. 30451922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7768055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triethyl 3-Methyl-4-phosphono-2-butenoate
CAS 41891-54-7
Molekylformel C11H21O5P
Kvantitet 5 g
Synonym Diethyl 3-ethoxycarbonyl-2-methyl-2-propenyl Phosphonate, 4-(Diethoxyphosphinyl)-3-methyl-2-butenoic acid Ethyl Ester, Triethyl 3-Methyl-4-phosphonocrotonate, Triethyl 4-Phosphono-3-methyl-2-butenoate
InChI formel InChI=1S/C11H21O5P/c1-5-14-11(12)8-10(4)9-17(13,15-6-2)16-7-3/h8H,5-7,9H2,1-4H3/b10-8+
LEDER CCOC(=O)\C=C(/C)\CP(=O)(OCC)OCC
IUPAC-namn ethyl (E)-4-diethoxyphosphoryl-3-methylbut-2-enoate
Molekylvikt (g/mol) 264.26
Formel vikt 264.1127