missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 4-phosphonocrotonate, cis + trans, 94%, Thermo Scientific Chemicals

Catalog Number p-7081148
Click to view available options
Quantity:
10 g
25 g
250 g
This item is not returnable. View return policy
10236-14-3
C10H19O5P
250.23
MFCD00009192
LXLODBXSCRTXFG-BQYQJAHWSA-N
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
5365783
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
CCOC(=O)\C=C\CP(=O)(OCC)OCC
This item is not returnable. View return policy
10236-14-3
C10H19O5P
250.23
MFCD00009192
LXLODBXSCRTXFG-BQYQJAHWSA-N
triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
5365783
ethyl (E)-4-diethoxyphosphorylbut-2-enoate
CCOC(=O)\C=C\CP(=O)(OCC)OCC

Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.13
Boiling Point 133°C to 135°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.455
Quantity 10 g
Formula Weight 250.23
Percent Purity 94%
Chemical Name or Material Triethyl 4-phosphonocrotonate, cis + trans

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'