Läs mer
Triethyl isocitrate, TRC
Produktkod. 30450274
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30450274 500 mg
1 options
Produktkod. 30450274 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT833990500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triethyl isocitrate
CAS 16496-37-0
Molekylformel C12 H20 O7
Kvantitet 500 mg
Synonym Triethyl isocitrate
InChI formel InChI=1S/C12H20O7/c1-4-17-9(13)7-8(11(15)18-5-2)10(14)12(16)19-6-3/h8,10,14H,4-7H2,1-3H3
LEDER CCOC(=O)CC(C(O)C(=O)OCC)C(=O)OCC
IUPAC-namn triethyl 1-hydroxypropane-1,2,3-tricarboxylate
Molekylvikt (g/mol) 276.283
Formel vikt 276.121