missing translation for 'onlineSavingsMsg'
Learn More

Triethyl methanetricarboxylate, 98%, Thermo Scientific Chemicals

Catalog Number p-4509974
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
6279-86-3
C10H16O6
232.23
AGZPNUZBDCYTBB-UHFFFAOYSA-N
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
80471
triethyl methanetricarboxylate
CCOC(=O)C(C(=O)OCC)C(=O)OCC
This item is not returnable. View return policy
6279-86-3
C10H16O6
232.23
AGZPNUZBDCYTBB-UHFFFAOYSA-N
tricarbethoxymethane, triethylmethanetricarboxylate, methanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-malonic acid diethyl ester, methanetricarboxylic acid triethyl ester, carboxymalonic acid triethyl ester, diethyl 2-ethoxycarbonyl malonate, diethyl 2-ethoxycarbonyl propane-1,3-dioate, pubchem3106, triethoxycarbonylmethane
80471
triethyl methanetricarboxylate
CCOC(=O)C(C(=O)OCC)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 29.0°C
Color Colorless to Yellow
Density 1.1000g/mL
Boiling Point 253.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH(CO2C2H5)3
Refractive Index 1.4240 to 1.4260
Quantity 25 mL
Beilstein 02,81
Specific Gravity 1.1
Solubility Information Solubility in water: immiscible
Formula Weight 232.23
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Triethyl methanetricarboxylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'