missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphonoacetate, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021687
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
867-13-0
C8H17O5P
224.193
MFCD00009177
GGUBFICZYGKNTD-UHFFFAOYSA-N
triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
13345
ethyl 2-diethoxyphosphorylacetate
CCOC(=O)CP(=O)(OCC)OCC
This item is not returnable. View return policy
867-13-0
C8H17O5P
224.193
MFCD00009177
GGUBFICZYGKNTD-UHFFFAOYSA-N
triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
13345
ethyl 2-diethoxyphosphorylacetate
CCOC(=O)CP(=O)(OCC)OCC

Triethyl phosphonoacetate serves as a reactant used in Horner-Wadsworth-Emmons reactions, Tsuji-Trost type reactions, Intramolecular Heck-type cyclization and isomerizations and Intramolecular aryne-ene reactions. In Horner-Wadsworth-Emmons reaction, it is utilized as a reagent to prepare chiral 2-methylcyclopropanecarboxylic acid from (S)-propylene oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphonoacetate serves as a reactant used in Horner-Wadsworth-Emmons reactions, Tsuji-Trost type reactions, Intramolecular Heck-type cyclization and isomerizations and Intramolecular aryne-ene reactions. In Horner-Wadsworth-Emmons reaction, it is utilized as a reagent to prepare chiral 2-methylcyclopropanecarboxylic acid from (S)-propylene oxide.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -24°C
Density 1.125
Boiling Point 260°C to 262°C
Flash Point 165°C (329°F)
Refractive Index 1.431
Quantity 100 g
Beilstein 1343714
Solubility Information Slightly miscible with water.
Formula Weight 224.19
Percent Purity ≥98%
Chemical Name or Material Triethyl phosphonoacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'