missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine trihydrofluoride, ca 37% HF, Thermo Scientific Chemicals

Catalog Number p-7046462
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
73602-61-6
C6H18F3N
161.212
MFCD00043294
IKGLACJFEHSFNN-UHFFFAOYSA-N
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3
175505
N,N-diethylethanamine;trihydrofluoride
CCN(CC)CC.F.F.F
This item is not returnable. View return policy
73602-61-6
C6H18F3N
161.212
MFCD00043294
IKGLACJFEHSFNN-UHFFFAOYSA-N
triethylamine trihydrofluoride, n,n-diethylethanamine trihydrofluoride, triethylammonium fluoride, ethanamine, n,n-diethyl-, trihydrofluoride, triethylamine trishydrofluoride, hydrogen fluoride in triethylamine, hf in triethylamine, triethylaminetrihydrofluoride, hydrogen fluoride triethylamine, ethanamine, n,n-diethyl-, hydrofluoride 1:3
175505
N,N-diethylethanamine;trihydrofluoride
CCN(CC)CC.F.F.F

Triethylamine trihydrofluoride acts as a mild and selective fluorinating agent used in the synthesis of acid fluorides and alkyl fluorides. It is also used in the synthesis of vicinal difluorides from epoxides, 3-fluoroazetidine. It is utilized as a selective reagent for the cleavage of O-silyl groups in carbohydrates, nucleotides and cyanohydrins. It is actively involved in the electrochemical fluorodeiodination of alkyl iodides and fluorodesilylation in beta-lactams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylamine trihydrofluoride acts as a mild and selective fluorinating agent used in the synthesis of acid fluorides and alkyl fluorides. It is also used in the synthesis of vicinal difluorides from epoxides, 3-fluoroazetidine. It is utilized as a selective reagent for the cleavage of O-silyl groups in carbohydrates, nucleotides and cyanohydrins. It is actively involved in the electrochemical fluorodeiodination of alkyl iodides and fluorodesilylation in beta-lactams.

Solubility
Miscible with water, chloroform and hexanes.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and glass.
TRUSTED_SUSTAINABILITY
Melting Point -29°C
Density 0.99
Boiling Point 70°C (15 mmHg)
Flash Point 87°C (188°F)
Linear Formula (CH3CH2)3N·3HF
Refractive Index 1.3915
Quantity 5 g
UN Number UN1790
Beilstein 5522945
Sensitivity Hygroscopic
Solubility Information Miscible with water,chloroform and hexanes.
Formula Weight 161.21
Assay ≈37% HF
Chemical Name or Material Triethylamine trihydrofluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'