Läs mer
Triethylenephosphoramide, TRC
Produktkod. 30134717
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134717 100 mg
18092410 25 mg
18031020 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134717 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT776600100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 545-55-1
Molekylformel C6 H12 N3 O P
Molekylvikt (g/mol) 173.15
Synonym Phosphine oxide, tris(1-aziridinyl)- (8CI), 1,1',1' '-Phosphinylidynetris[aziridine], APO, Aphoxide, ENT 24915, Imperon Fixer T, NSC 9717, Phosphoric acid triethylene imide, Phosphoric triamide, N,N',N' '-tri-1,2-ethanediyl-, TAPO, TEF, TEPA, TEPA (pesticide), Tri-1-aziridinylphosphine oxide, Triaziridinophosphine oxide, Triaziridinylphosphine oxide, Triethylenephosphoramide, Triethylenephosphorotriamide, Tris(1-aziridine)phosphine oxide, Tris(1-aziridinyl)phosphine oxide, Tris(aziridinyl)phosphine oxide
IUPAC-namn 1-[bis(aziridin-1-yl)phosphoryl]aziridine
LEDER O=P(N1CC1)(N2CC2)N3CC3
Specifikationer
Kemiskt namn eller material Triethylenephosphoramide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C6H12N3OP/c10-11(7-1-2-7,8-3-4-8)9-5-6-9/h1-6H2
Formel vikt 173.0718
Procent renhet >95%