Läs mer
Triethyloxonium Tetrachloroferrate(III), TRC
Produktkod. 30455926
Change view
Kvantitet:
2.5 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455926 2.5 g
30450899 5 g
30454948 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7769052.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15555-35-8
Molekylformel C6H15Cl4FeO
Molekylvikt (g/mol) 300.84
Synonym Triethyloxonium Chloroferrate, Triethyloxonium Tetrachloroferrate
IUPAC-namn tetrachloroiron(1+);triethyloxidanium
LEDER CC[O+](CC)CC.Cl[Fe+](Cl)(Cl)Cl
Specifikationer
Kemiskt namn eller material Triethyloxonium Tetrachloroferrate(III)
Kvantitet 2.5 g
InChI formel InChI=1S/C6H15O.4ClH.Fe/c1-4-7(5-2)6-3;;;;;/h4-6H2,1-3H3;4*1H;/q+1;;;;;+5/p-4
Formel vikt 298.9226