missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium iodide, 97%, Thermo Scientific Chemicals

Catalog Number p-7043412
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1829-92-1
C6H15IS
246.15
MFCD00051867
LKDQWVKWYGOVJW-UHFFFAOYSA-M
triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide
74589
triethylsulfanium;iodide
CC[S+](CC)CC.[I-]
This item is not returnable. View return policy
1829-92-1
C6H15IS
246.15
MFCD00051867
LKDQWVKWYGOVJW-UHFFFAOYSA-M
triethylsulfonium iodide, triethylsulfanium iodide, triethylsulphonium iodide, sulfonium, triethyl-, iodide, acmc-20aje7, sulfonium,triethyl-,iodide
74589
triethylsulfanium;iodide
CC[S+](CC)CC.[I-]

Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsulfonium iodide is used as an ethylidene-transfer reagent. It is used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Light, Heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY
Melting Point ∽148°C (decomposition)
Quantity 5 g
Beilstein 3911790
Sensitivity Light sensitive
Solubility Information It is soluble in water.
Formula Weight 246.15
Percent Purity 97%
Chemical Name or Material Triethylsulfonium iodide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'