Läs mer
Invitrogen™ TRIF Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595104
Beskrivning
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Involved in innate immunity against invading pathogens. Adapter used by TLR3 and TLR4 (through TICAM2) to mediate NF-kappa-B and interferon-regulatory factor (IRF) activation, and to induce apoptosis. Ligand binding to these receptors results in TRIF recruitment through its TIR domain. Distinct protein-interaction motifs allow recruitment of the effector proteins TBK1, TRAF6 and RIPK1, which in turn, lead to the activation of transcription factors IRF3 and IRF7, NF-kappa-B and FADD respectively.
Specifikationer
| TRIF | |
| Polyclonal | |
| Unconjugated | |
| TICAM1 | |
| AW046014; AW547018; hCG_39321; HSD34; IIAE6; MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB; Putative NF-kappa-B-activating protein 502H; RNF36; TI; Ticam1; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; TIR domain-containing adaptor inducing interferon-beta; TIR-domain containing adaptor inducing interferon-beta; toll like receptor adaptor molecule 1; toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta; toll-interleukin I receptor domain containing adaptor molecule 1; toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1; TRIF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 106759 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q80UF7 | |
| TICAM1 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of mouse TRIF (693-708aa NNHMWGHTGAQSSDDK). | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.