Läs mer
Trifloxysulfuron Sodium Salt, TRC
Produktkod. 30449813
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30449813 50 mg
30452845 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30449813 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT77900050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 199119-58-9
Molekylformel C14 H13 F3 N5 O6 S . Na
Molekylvikt (g/mol) 459.33
Synonym 2-Pyridinesulfonamide, N-[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]-3-(2,2,2-trifluoroethoxy)-, monosodium salt (9CI), CGA 362622, Envoke, Monument, NOJ 120, Sodium trifloxysulfuron, Trifloxysulfuron sodium salt, Trifloxysulfuron-sodium
IUPAC-namn sodium;(4,6-dimethoxypyrimidin-2-yl)carbamoyl-[3-(2,2,2-trifluoroethoxy)pyridin-2-yl]sulfonylazanide
LEDER [Na+].COc1cc(OC)nc(NC(=O)[N-]S(=O)(=O)c2ncccc2OCC(F)(F)F)n1
Specifikationer
Kemiskt namn eller material Trifloxysulfuron Sodium Salt
Kvantitet 50 mg
InChI formel InChI=1S/C14H14F3N5O6S.Na/c1-26-9-6-10(27-2)20-12(19-9)21-13(23)22-29(24,25)11-8(4-3-5-18-11)28-7-14(15,16)17;/h3-6H,7H2,1-2H3,(H2,19,20,21,22,23);/q;+1/p-1
Formel vikt 459.0436