Läs mer
Trifluoperazine Sulfone, Mikromol™
Produktkod. 30049918
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049918 25 mg
30049796 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049918 Leverantör Mikromol Leverantörsnummer MM0441.040025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Trifluoperazine Sulfone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trifluoperazine Sulfone
CAS 2230802-07-8
Analyt- eller komponentnamn Trifluoperazine Sulfone
Frakt skick Room Temperature
Molekylformel C21 H24 F3 N3 O2 S
Certifikat och efterlevnad ISO 9001
Synonym Trifluoperazine Sulfone
InChI formel InChI=1S/C21H24F3N3O2S/c1-25-11-13-26(14-12-25)9-4-10-27-17-5-2-3-6-19(17)30(28,29)20-8-7-16(15-18(20)27)21(22,23)24/h2-3,5-8,15H,4,9-14H2,1H3
LEDER CN1CCN(CCCN2c3ccccc3S(=O)(=O)c4ccc(cc24)C(F)(F)F)CC1
IUPAC-namn 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine 5,5-dioxide
Molekylvikt (g/mol) 439.49
Kvantitet 25 mg
Formel vikt 439.1541 g/mol