missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391495
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
76-05-1
C2HF3O2
114.02
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
6422
CHEBI:45892
trifluoroacetic acid
OC(=O)C(F)(F)F
This item is not returnable. View return policy
76-05-1
C2HF3O2
114.02
MFCD00004169
DTQVDTLACAAQTR-UHFFFAOYSA-N
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
6422
CHEBI:45892
trifluoroacetic acid
OC(=O)C(F)(F)F

High quality chemicals for laboratory and specialized industrial use

Formula Weight 114.03
Grade BAKER ANALYZED™ Reagent
Quantity 500 mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.53kg
Chemical Name or Material Trifluoroacetic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'