Läs mer
Trifluoromethylsulfonamide, TRC
Produktkod. 30449475
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30449475 5 g
1 options
Produktkod. 30449475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7924105G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trifluoromethylsulfonamide
CAS 421-85-2
Molekylformel CH2F3NO2S
Kvantitet 5 g
Synonym 1,1,1-Trifluoromethanesulfonamide, Trifluoromethanesulfonamide
InChI formel InChI=1S/CH2F3NO2S/c2-1(3,4)8(5,6)7/h(H2,5,6,7)
LEDER NS(=O)(=O)C(F)(F)F
IUPAC-namn trifluoromethanesulfonamide
Molekylvikt (g/mol) 149.09
Formel vikt 148.9758