Läs mer
Triflusulfuron, TRC
Produktkod. 30134413
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134413 100 mg
30009131 25 mg
30007619 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134413 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT782005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 135990-29-3
Molekylformel C16H17F3N6O6S
Molekylvikt (g/mol) 478.4
Synonym 2-[[[[[4-(Dimethylamino)-6-(2,2,2-trifluoroethoxy)-1,3,5-triazin-2-yl]amino]carbonyl]amino]sulfonyl]-3-methylbenzoic Acid, Debut;
IUPAC-namn 2-[[4-(dimethylamino)-6-(2,2,2-trifluoroethoxy)-1,3,5-triazin-2-yl]carbamoylsulfamoyl]-3-methylbenzoic acid
LEDER O=C(O)C1=C(S(NC(NC2=NC(N(C)C)=NC(OCC(F)(F)F)=N2)=O)(=O)=O)C(C)=CC=C1
Specifikationer
Kemiskt namn eller material Triflusulfuron
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C16H17F3N6O6S/c1-8-5-4-6-9(11(26)27)10(8)32(29,30)24-14(28)21-12-20-13(25(2)3)23-15(22-12)31-7-16(17,18)19/h4-6H,7H2,1-3H3,(H,26,27)(H2,20,21,22,23,24,28)
Formel vikt 478.0882
Procent renhet >95%