Läs mer
Trihexyl O-Butyrylcitrate, TRC
Produktkod. 30449871
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30449871 25 g
1 options
Produktkod. 30449871 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT93571025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trihexyl butyryl citrate
CAS 82469-79-2
Molekylformel C28 H50 O8
Kvantitet 25 g
Synonym 1,2,3-Propanetricarboxylic acid','2-(1-oxobutoxy)-','1,2,3-trihexyl ester, 1,2,3-Propanetricarboxylic acid','2-(1-oxobutoxy)-','trihexyl ester (9CI), Butyryl tri-n-hexyl citrate, Butyryl trihexyl citrate, Citroflex B 6, Trihexyl butyrylcitrate, Trihexyl citrate butyrate, Plastic additive 25, Citric Acid','Butyl Trihexyl Ester
InChI formel InChI=1S/C28H50O8/c1-5-9-12-15-19-33-25(30)22-28(36-24(29)18-8-4,27(32)35-21-17-14-11-7-3)23-26(31)34-20-16-13-10-6-2/h5-23H2,1-4H3
LEDER CCCCCCOC(=O)CC(CC(=O)OCCCCCC)(OC(=O)CCC)C(=O)OCCCCCC
IUPAC-namn trihexyl 2-butanoyloxypropane-1,2,3-tricarboxylate
Molekylvikt (g/mol) 514.69
Formel vikt 514.3506