missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TRIM25 (Human) Recombinant Protein
Human TRIM25 partial ORF ( NP_005073, 211 a.a. - 310 a.a.) recombinant protein with GST-tag at N-terminal.
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005073 |
---|---|
Gene ID (Entrez) | 7706 |
Name | tripartite motif-containing 25 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16107855
|
Abnova™
H00007706-Q01.10UG |
10 μg |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16117855
|
Abnova™
H00007706-Q01.25UG |
25 μg |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: SQINGASRALDDVRNRQQDVRMTANRKVEQLQQEYTEMKALLDASETTSTRKIKEEEKRVNSKFDTIYQILLKKKSEIQTLKEEIEQSLTKRDEFEFLEK
Specifications
NP_005073 | |
tripartite motif-containing 25 | |
125% SDS-PAGE Stained with Coomassie Blue | |
EFP, RNF147, Z147, ZNF147 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
7706 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TRIM25 | |
GST |