missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ TRIM36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528401
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: A549, HCT116. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifikationer
| TRIM36 | |
| Polyclonal | |
| Unconjugated | |
| TRIM36 | |
| Acrosome RBCC protein; D18Wsu100e; E3 ubiquitin-protein ligase Trim36; haprin; RBCC728; RING finger protein 98; RING-type E3 ubiquitin transferase TRIM36; RNF98; trim36; tripartite motif containing 36; tripartite motif protein 36; tripartite motif-containing 36; tripartite motif-containing protein 36; wu:fj48f01; zgc:153447; zgc:63476; Zinc-binding protein Rbcc728 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55521 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NQ86 | |
| TRIM36 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human TRIM36. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering