missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TRIM41 Antibody Blocking Peptide
Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Kvantitet:
0,1 mg
Specifications
Specifications
| Gen-ID (Entrez) | 90933 |
| Koncentration | 1 mg/mL |
| Innehåll och lagring | Store at -80°C. Avoid freeze/thaw cycles. |
| För användning med (applikation) | This Peptide is Useful as a Blocking Peptide for NBP3-05293. |
| Gene Alias | tripartite motif containing 41 |
| Gensymbol | TRIM41 |
| Kvantitet | 0,1 mg |
| Forskningskategori | Autophagy |
| Förvaringsbuffert | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction