Läs mer
Trimetazidine Dihydrochloride, TRC
Produktkod. 30453495
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30453495 1 g
1 options
Produktkod. 30453495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7956101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trimetazidine Dihydrochloride
CAS 13171-25-0
Anteckningar om renhetsgrad HPLC
Molekylformel C14 H22 N2 O3 . 2 Cl H
Kvantitet 1 g
Synonym Trimetazidine Dihydrochloride, Piperazine','1-[(2,3,4-trimethoxyphenyl)methyl]-','dihydrochloride (9CI), Piperazine','1-(2,3,4-trimethoxybenzyl)-','dihydrochloride (7CI,8CI), 1-(2,3,4-Trimethoxybenzyl)piperazine dihydrochloride, Kyurinett, S 4004, Vastarel, Vastarel F, Yoshimilon
InChI formel InChI=1S/C14H22N2O3.2ClH/c1-17-12-5-4-11(13(18-2)14(12)19-3)10-16-8-6-15-7-9-16;;/h4-5,15H,6-10H2,1-3H3;2*1H
LEDER Cl.Cl.COc1ccc(CN2CCNCC2)c(OC)c1OC
IUPAC-namn 1-[(2,3,4-trimethoxyphenyl)methyl]piperazine;dihydrochloride
Molekylvikt (g/mol) 339.26
Formel vikt 338.1164