Läs mer
Trimethoxy(2-phenylethyl)silane, TRC
Produktkod. 30454485
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30454485 1 g
1 options
Produktkod. 30454485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7974551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trimethoxy(2-phenylethyl)silane
CAS 49539-88-0
Molekylformel C11H18O3Si
Kvantitet 1 g
Synonym [2-(Trimethoxysilyl)ethyl]benzene, (2-Phenylethyl)trimethoxysilane, P 113, P 113 (Silane Derivative), Phenethyltrimethoxysilane, SIP 6722.6, T 2869, β-(Trimethoxysilyl)ethylbenzene
InChI formel InChI=1S/C11H18O3Si/c1-12-15(13-2,14-3)10-9-11-7-5-4-6-8-11/h4-8H,9-10H2,1-3H3
LEDER CO[Si](CCC1=CC=CC=C1)(OC)OC
IUPAC-namn trimethoxy(phenethyl)silane
Molekylvikt (g/mol) 226.34
Formel vikt 226.1