Läs mer
Trimethyl Alpha-(Acetamido)phosphonoacetate, TRC
Produktkod. 30471494
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30471494 50 mg
1 options
Produktkod. 30471494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM21925050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trimethyl alpha-(Acetamido)phosphonoacetate
CAS 89524-99-2
Molekylformel C7H14NO6P
Kvantitet 50 mg
Synonym Methyl 2-(Acetylamino)-2-(dimethoxyphosphinyl)acetate, Methyl alpha-(Acetylamino)-alpha-(dimethoxyphosphoryl)acetate
InChI formel InChI=1S/C7H14NO6P/c1-5(9)8-6(7(10)12-2)15(11,13-3)14-4/h6H,1-4H3,(H,8,9)
LEDER COC(=O)C(NC(=O)C)P(=O)(OC)OC
IUPAC-namn methyl 2-acetamido-2-dimethoxyphosphorylacetate
Molekylvikt (g/mol) 239.16
Formel vikt 239.0559