Läs mer
Trimethyl[(tributylstannyl)ethynyl]silane, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Trimethyl[(tributylstannyl)ethynyl]silane |
| CAS | 81353-38-0 |
| Molekylformel | C17H36SiSn |
| Kvantitet | 5 g |
| Synonym | Tributylstannyl)(trimethylsilyl)acetylene, (Trimethylsilylethynyl)tributylstannane, Tributyl(trimethylsilylethynyl)stannane, Tributyl(trimethylsilylethynyl)tin, Trimethyl(2-tributylstannylethynyl)silan, Trimethyl[(tributylstannyl)ethynyl]silane |
| InChI formel | InChI=1S/C5H9Si.3C4H9.Sn/c1-5-6(2,3)4;3*1-3-4-2;/h2-4H3;3*1,3-4H2,2H3; |
| LEDER | CCCC[Sn](CCCC)(CCCC)C#C[Si](C)(C)C |
| IUPAC-namn | trimethyl(2-tributylstannylethynyl)silane |
| Molekylvikt (g/mol) | 387.26 |
| Formel vikt | 388.1608 |