Läs mer
Trimethylene Di(thiotosylate), TRC
Produktkod. 30456332
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30456332 5 g
1 options
Produktkod. 30456332 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7961835G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trimethylene Di(thiotosylate)
CAS 3866-79-3
Molekylformel C17H20O4S4
Kvantitet 5 g
Synonym 4-Methyl-benzenesulfonothioic Acid S,S'-1,3-Propanediyl Ester, 1,3-Bis(4-methylbenzenesulfonylthio)propane, 1,3-Propanedithiol Ditosylate, NSC 82472, Trimethylene di-p-Toluenethiosulfonate
InChI formel InChI=1S/C17H20O4S4/c1-14-4-8-16(9-5-14)24(18,19)22-12-3-13-23-25(20,21)17-10-6-15(2)7-11-17/h4-11H,3,12-13H2,1-2H3
LEDER Cc1ccc(cc1)S(=O)(=O)SCCCSS(=O)(=O)c2ccc(C)cc2
IUPAC-namn 1-methyl-4-[3-(4-methylphenyl)sulfonylsulfanylpropylsulfanylsulfonyl]benzene
Molekylvikt (g/mol) 416.6
Formel vikt 416.0244