Läs mer
Trimethylolpropane Tris(2-methyl-1-aziridinepropionate), TRC
Produktkod. 30456664
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30456664 250 mg
1 options
Produktkod. 30456664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT796975250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trimethylolpropane Tris(2-methyl-1-aziridinepropionate)
CAS 64265-57-2
Molekylformel C24H41N3O6
Kvantitet 250 mg
Synonym 2-Ethyl-2-[[3-(2-methyl-1-aziridinyl)-1-oxopropoxy]methyl]-1,3-propanediyl Bis(2-methyl-1-aziridinepropanoate), AS 316, CX 100, Crosslinker CX 100, Crosslinker SC 100, Ionac PFAZ 322, NeoCryl CX 100, PFAZ 322, PZ 28, Permatex XR 2500, Trimethylolpropane Tris(2-methyl-1-aziridinyl)propionate, XC 113, XR 2500
InChI formel InChI=1S/C24H41N3O6/c1-5-24(15-31-21(28)6-9-25-12-18(25)2,16-32-22(29)7-10-26-13-19(26)3)17-33-23(30)8-11-27-14-20(27)4/h18-20H,5-17H2,1-4H3
LEDER CCC(COC(=O)CCN1CC1C)(COC(=O)CCN2CC2C)COC(=O)CCN3CC3C
IUPAC-namn 2,2-bis[3-(2-methylaziridin-1-yl)propanoyloxymethyl]butyl 3-(2-methylaziridin-1-yl)propanoate
Molekylvikt (g/mol) 467.6
Formel vikt 467.2995