missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyl azide, 94%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.
Solubility
Miscible with toluene, dichloromethane, diethyl ether and most organic solvents.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
Trimethylsilyl azide is used as an azidonation reagent for amines, amides, aldehydes and ketones. It is involved in the preparation of alfa- and beta-siloxy azides from carbonyl compounds and epoxides. It is also used in the preparation of heterocyclic compounds and also acts as an effective substitute for hydrazoic acid. Further, it is used in the Oseltamivir total synthesis.
Solubility
Miscible with toluene, dichloromethane, diethyl ether and most organic solvents.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | -95°C |
Density | 0.872 |
Boiling Point | 95°C to 99°C |
Flash Point | 6°C (42°F) |
Refractive Index | 1.414 |
Quantity | 5 g |
UN Number | UN1992 |
Beilstein | 1903730 |
Sensitivity | Moisture sensitive |
Solubility Information | Miscible with toluene,dichloromethane,diethyl ether and most organic solvents. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'