missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin hydroxide, 98%, Thermo Scientific Chemicals

Catalog Number p-4813698
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
56-24-6
C3H10OSn
180.82
MFCD00013929
OJZNYUDKNVNEMV-UHFFFAOYSA-M
trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide
6328620
trimethyltin;hydrate
C[Sn](C)(C)O
56-24-6
C3H10OSn
180.82
MFCD00013929
OJZNYUDKNVNEMV-UHFFFAOYSA-M
trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide
6328620
trimethyltin;hydrate
C[Sn](C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 118.0°C to 119.0°C
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3SnOH
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 180.82
Percent Purity 98%
Chemical Name or Material Trimethyltin hydroxide
compliance-icons compliance-icons
Product Identifier
  • Trimethyltin hydroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'