missing translation for 'onlineSavingsMsg'
Learn More

Trimethyltin hydroxide, 98%, ACROS Organics™

Brand:  Acros Organics 449220010

Additional Details : CAS Number : 56-24-6 Weight : 0.00100kg Transport : UN number : 3146 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 1g

 View more versions of this product

Product Code. 15214764

  • 610.33 SEK / 1g


In stock, ready to ship: 9
Add to basket


Specifications

Chemical Identifiers

56-24-6
180.82
trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide
trimethyltin;hydrate
C3H10OSn
UABRYPURASNHRO-UHFFFAOYSA-N
6328620
C[Sn](C)C.O

Specifications

Trimethyltin hydroxide
118.0°C to 119.0°C
56-24-6
C3H10OSn
trimethyltin hydroxide, stannane, hydroxytrimethyl, trimethylstannanol, hydroxytrimethyltin, trimethyl tin hydroxide, trimethylhydroxytin, hydroxytrimethylstannane, trimethylstannyl hydroxide, unii-10alt8dj2v, tin, trimethyl-, hydroxide
UABRYPURASNHRO-UHFFFAOYSA-N
trimethyltin;hydrate
6328620
98%
Glass bottle
1g
98%
(CH3)3SnOH
Solubility in water: soluble
C[Sn](C)C.O
180.82
180.82
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trimethyltin hydroxide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1

Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Documents