Läs mer
Trioctyl Trimellitate, TRC
Produktkod. 30453741
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30453741 10 g
1 options
Produktkod. 30453741 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT80440010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tri(2-ethylhexyl) trimellitat
CAS 3319-31-1
Molekylformel C33 H54 O6
Kvantitet 10 g
Synonym 1,2,4-Benzenetricarboxylic acid','tris(2-ethylhexyl) ester (7CI,8CI,9CI), ADK Cizer TOTM, Adekacizer TOTM, Bisoflex TOT, Codan Set 43.4400, Diplast TM/ST, Eastman TOTM, Hatcol 200, Kodaflex TOTM, Monocizer W 700, Monocizer W 705, Monocizer W 710L, Morflex 510, Nuoplaz 6959, Octyl trimellitate, Pelemol TOTM, Plasthall TOTM, Reomol OTM, Sansocizer TOTM, Staflex TOTM, T 08, TM 150, TOTM, TOTM-I, TOTM-NB, Tri-2-ethylhexyl trimellitate, Trimex T 08, Trioctyl 1,2,4-benzenetricarboxylate, Trioctyl trimellitate, Tris(2-ethylhexyl) trimellitate, Tris(octyl) trimellitate, Uniplex 546, Uniplex 546A, W 700, W 705
InChI formel InChI=1S/C33H54O6/c1-7-13-16-25(10-4)22-37-31(34)28-19-20-29(32(35)38-23-26(11-5)17-14-8-2)30(21-28)33(36)39-24-27(12-6)18-15-9-3/h19-21,25-27H,7-18,22-24H2,1-6H3
LEDER CCCCC(CC)COC(=O)c1ccc(C(=O)OCC(CC)CCCC)c(c1)C(=O)OCC(CC)CCCC
IUPAC-namn tris(2-ethylhexyl) benzene-1,2,4-tricarboxylate
Molekylvikt (g/mol) 546.7783
Formel vikt 546.392