missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIP6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21163-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the zyxin family and encodes a protein with three LIM zinc-binding domains. This protein localizes to focal adhesion sites and along actin stress fibers. Recruitment of this protein to the plasma membrane occurs in a lysophosphatidic acid (LPA)-dependent manner and it regulates LPA-induced cell migration. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.Specifications
TRIP6 | |
Polyclonal | |
Unconjugated | |
TRIP6 | |
OIP 1, OIP1, Opa interacting protein 1, TR interacting protein 6, TRIP 6, TRIP6, ZRP 1, Zyxin related protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22051, 686323, 7205 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15654, Q9Z1Y4 | |
TRIP6 | |
TRIP6 Fusion Protein Ag15149 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |