Läs mer
Tripelennamine Hydrochloride, TRC
Produktkod. 30453951
Change view
Kvantitet:
50 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453951 50 mg
30451338 10 g
30453435 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30453951 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT80832550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154-69-8
Molekylformel C16 H21 N3 . Cl H
Molekylvikt (g/mol) 291.82
Synonym 1,2-Ethanediamine, N,N-dimethyl-N'-(phenylmethyl)-N'-2-pyridinyl-, monohydrochloride, 1,2-Ethanediamine, N1,N1-dimethyl-N2-(phenylmethyl)-N2-2-pyridinyl-, hydrochloride, Pyridine, 2-[benzyl[2-(dimethylamino)ethyl]amino]-, monohydrochloride, 2-[Benzyl[2-(dimethylamino)ethyl]amino]pyridine hydrochloride, Azaron, Dehistin, Dehistin monohydrochloride, N,N-Dimethyl-N'-(2-pyridyl)-N'-benzylethylenediamine hydrochloride, N-Benzyl-N',N'-dimethyl-N-2-pyridylethylenediamine hydrochloride, NIH 10186, PBZ, Piristin, Pyribenzamine hydrochloride, Pyribenzamine monohydrochloride, Pyrinamine, ReCovr, Stanzamine, Tripelennamine hydrochloride, Tripelennamine monohydrochloride, Vetibenzamine, Vetobenzamina
IUPAC-namn N'-benzyl-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
LEDER Cl.CN(C)CCN(Cc1ccccc1)c2ccccn2
Specifikationer
Kemiskt namn eller material Tripelennamine hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C16H21N3.ClH/c1-18(2)12-13-19(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15;/h3-11H,12-14H2,1-2H3;1H
Formel vikt 291.1502