Läs mer
Tripelennamine N-Oxide, Mikromol™
Produktkod. 30049565
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049565 100 mg
30050348 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049565 Leverantör Mikromol Leverantörsnummer MM1631.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Tripelennamine N-Oxide
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tripelennamine N-Oxide
CAS 60317-19-3
Analyt- eller komponentnamn Tripelennamine N-Oxide
Frakt skick Room Temperature
Molekylformel C16 H21 N3 O
Certifikat och efterlevnad ISO 9001
Synonym 1,2-Ethanediamine, N1,N1-dimethyl-N2-(phenylmethyl)-N2-2-pyridinyl-, N1-oxide, 1,2-Ethanediamine, N,N-dimethyl-N'-(phenylmethyl)-N'-2-pyridinyl-, N-oxide (9CI), Tripelennamine, N-oxide (6CI)
InChI formel InChI=1S/C16H21N3O/c1-19(2,20)13-12-18(16-10-6-7-11-17-16)14-15-8-4-3-5-9-15/h3-11H,12-14H2,1-2H3
LEDER C[N+](C)([O-])CCN(Cc1ccccc1)c2ccccn2
IUPAC-namn 2-[benzyl(pyridin-2-yl)amino]-N,N-dimethylethanamine oxide
Molekylvikt (g/mol) 271.36
Kvantitet 100 mg
Formel vikt 271.1685 g/mol