Läs mer
Triphenyl(p-carboxyphenyl)phosphonium Chloride, TRC
Produktkod. 30413950
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30413950 25 mg
1 options
Produktkod. 30413950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT27451025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triphenyl(p-carboxyphenyl)phosphonium Chloride
CAS 35812-45-4
Molekylformel C25H20O2PCl
Kvantitet 25 mg
Synonym Phosphonium','(4-carboxyphenyl)triphenyl-','chloride (9CI), (p-Carboxyphenyl)triphenylphosphonium chloride, Triphenyl(p-carboxyphenyl)phosphonium chloride
InChI formel InChI=1S/C25H19O2P.ClH/c26-25(27)20-16-18-24(19-17-20)28(21-10-4-1-5-11-21,22-12-6-2-7-13-22)23-14-8-3-9-15-23;/h1-19H;1H
LEDER [Cl-].O=C(O)C1=CC=C(C=C1)[P+](C=2C=CC=CC2)(C=3C=CC=CC3)C=4C=CC=CC4
IUPAC-namn (4-carboxyphenyl)triphenylphosphonium chloride
Molekylvikt (g/mol) 418.86
Formel vikt 418.09