Läs mer
Triphenylacetonitrile, TRC
Produktkod. 30449270
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30449270 100 mg
1 options
Produktkod. 30449270 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT394360100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triphenylacetonitrile
CAS 6639-43-6
Molekylformel C20 H15 N
Kvantitet 100 mg
Synonym Triphenyl-acetonitrile, α,α-Diphenylbenzeneacetonitrile, Cyanotriphenylmethane, NSC 16078, Triphenylmethyl cyanide, Trityl cyanide, 2,2,2-Triphenylacetonitrile
InChI formel InChI=1S/C20H15N/c21-16-20(17-10-4-1-5-11-17,18-12-6-2-7-13-18)19-14-8-3-9-15-19/h1-15H
LEDER N#CC(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC-namn 2,2,2-triphenylacetonitrile
Molekylvikt (g/mol) 269.34
Formel vikt 269.12