Läs mer
Triphenylsilanethiol, TRC
Produktkod. 30418414
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30418414 100 mg
1 options
Produktkod. 30418414 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP336840100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triphenylsilanethiol
CAS 14606-42-9
Molekylformel C18 H16 S Si
Kvantitet 100 mg
Synonym 1,1,1-triphenyl-Silanethiol, triphenyl-Silanethiol, 1,1,1-Triphenylsilanethiol, Mercaptotriphenylsilane, Triphenylsilyl mercaptan
InChI formel InChI=1S/C18H16SSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15,19H
LEDER S[Si](c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC-namn triphenyl(sulfanyl)silane
Molekylvikt (g/mol) 292.47
Formel vikt 292.074