Läs mer
Triphenylsilylamine, TRC
Produktkod. 30455820
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455820 250 mg
30452639 100 mg
30450168 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455820 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT809085250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4215-80-9
Molekylformel C18H17NSi
Molekylvikt (g/mol) 275.42
Synonym 1,1,1-Triphenylsilanamine, 1,1,1-Triphenylsilylamine, Aminotriphenylsilane
IUPAC-namn [amino(diphenyl)silyl]benzene
LEDER N[Si](c1ccccc1)(c2ccccc2)c3ccccc3
Specifikationer
Kemiskt namn eller material Triphenylsilylamine
Kvantitet 250 mg
InChI formel InChI=1S/C18H17NSi/c19-20(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H,19H2
Formel vikt 275.113