Läs mer
Triptolide O-Methyl Phosphate Dibenzyl Ester, TRC
Produktkod. 30454540
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30454540 5 mg
1 options
Produktkod. 30454540 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8156255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Triptolide O-Methyl Phosphate Dibenzyl Ester
CAS 1254702-92-5
Molekylformel C35H39O10P
Kvantitet 5 mg
Synonym Minnelide Dibenzyl Ester, [[(3bS,4aS,5aR,6R,6aS,7aS,7bS,8aS,8bS)-1,3,3b,4,4a,6,6a,7a,7b,8b,9,10-Dodecahydro-8b-methyl-6a-(1-methylethyl)-1-oxotrisoxireno[4b,5:6,7:8a,9]phenanthro[1,2-c]furan-6-yl]oxy]methyl Phosphoric Acid Bis(phenylmethyl) Ester
InChI formel InChI=1S/C35H39O10P/c1-21(2)33-28(44-33)29-35(45-29)32(3)15-14-24-25(19-38-30(24)36)26(32)16-27-34(35,43-27)31(33)39-20-42-46(37,40-17-22-10-6-4-7-11-22)41-18-23-12-8-5-9-13-23/h4-13,21,26-29,31H,14-20H2,1-3H3/t26-,27-,28-,29-,31+,32-,33-,34+,35+/m0/s1
LEDER [H][C@]12C3=C(C(OC3)=O)CC[C@]1(C)[C@@]45[C@@]([C@H](OCOP(OCC6=CC=CC=C6)(OCC7=CC=CC=C7)=O)C(C(C)C)(O8)[C@@H]8[C@@H]4O5)(O9)[C@@H]9C2
Molekylvikt (g/mol) 650.65
Formel vikt 650.2281