Läs mer
Tris(1-chloro-2-propyl) Phosphate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Tris-(2-chloropropyl)phosphate |
| CAS | 13674-84-5 |
| Molekylformel | C9 H18 Cl3 O4 P |
| Kvantitet | 250 mg |
| Synonym | 2-Propanol','1-chloro-','2,2',2''-phosphate, 2-Propanol','1-chloro-','phosphate (3:1) (8CI,9CI), Amgard TMCP, Antiblaze 80, Antiblaze TMCP, Daltoguard F, Fyrol PCF, Hostaflam OP 820, Levagard PP, Levagard PP-Z, Levagard TCPP, PUMA 4010, Phireguard TCPP, TCPP, Tolgard TMCP, Tris(1-chloro-2-propyl) phosphate, Tris(1-methyl-2-chloroethyl) phosphate, Tris(2-chloro-1-methylethyl) phosphate, Tris(2-chloroisopropyl) phosphate, Tris(chloroisopropyl) phosphate, Tris(β-chloroisopropyl) phosphate, WSFR-TCPP, Yoke TCPP |
| InChI formel | InChI=1S/C9H18Cl3O4P/c1-7(4-10)14-17(13,15-8(2)5-11)16-9(3)6-12/h7-9H,4-6H2,1-3H3 |
| LEDER | CC(CCl)OP(=O)(OC(C)CCl)OC(C)CCl |
| IUPAC-namn | tris(1-chloropropan-2-yl) phosphate |
| Molekylvikt (g/mol) | 327.57 |
| Formel vikt | 326.0008 |