Läs mer
Tris(2,2,2-trifluoroethyl)orthoformate, TRC
Produktkod. 30451149
Change view
Kvantitet:
50 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30451149 500 mg
30454364 50 mg
30453576 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30451149 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT876723500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58244-27-2
Molekylformel C7H7F9O3
Molekylvikt (g/mol) 310.11
Synonym 2-[bis(2,2,2-Trifluoroethoxy)methoxy]-1,1,1-trifluoroethane
IUPAC-namn 2-[bis(2,2,2-trifluoroethoxy)methoxy]-1,1,1-trifluoroethane
LEDER FC(F)(F)COC(OCC(F)(F)F)OCC(F)(F)F
Specifikationer
Kemiskt namn eller material Tris(2,2,2-trifluoroethyl)orthoformate
Kvantitet 500 mg
InChI formel InChI=1S/C7H7F9O3/c8-5(9,10)1-17-4(18-2-6(11,12)13)19-3-7(14,15)16/h4H,1-3H2
Formel vikt 310.0251