missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Tris(2-chloroethyl) phosphate, Dr. Ehrenstorfer
Specifications
Specifications
| Kemiskt namn eller material | Tris-(2-chloroethyl)phosphate |
| CAS | 115-96-8 |
| Analyt- eller komponentnamn | Tris-(2-chloroethyl)phosphate |
| Frakt skick | Room Temperature |
| Molekylformel | C6 H12 Cl3 O4 P |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | 2-Chloroethanol Phosphate, 3CF, Amgard TCEP, CLP, Celluflex CEF, Disflamoll TCA, Fyrol CEF, Fyrol CF, Genomoll P, NSC 3213, Niax 3CF, Tri(chloroethyl) Phosphate, Tri(β-chloroethyl)phosphate, Tris(2-chloroethyl)orthophosphate, Tri(2-chloroethyl)phosphate, Tris(β-chloroethyl)phosphate; |
| InChI formel | InChI=1S/C6H12Cl3O4P/c7-1-4-11-14(10,12-5-2-8)13-6-3-9/h1-6H2 |
| Lösningstyp | Neat |
| LEDER | ClCCOP(=O)(OCCCl)OCCCl |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?