Läs mer
Tris(2-methoxyethoxy)vinylsilane, 98%, Thermo Scientific™
Produktkod. 15417208
Change view
Kvantitet:
100 mL
500 mL
Förpackningsstorlek:
100ml
500ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15417208 100 mL 100ml
15427208 500 mL 500ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15417208 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H60717.AE
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1067-53-4
Molekylformel C11H24O6Si
Molekylvikt (g/mol) 280.392
MDL-nummer MFCD00008500
InChI-nyckel WOXXJEVNDJOOLV-UHFFFAOYSA-N
Synonym tris 2-methoxyethoxy vinylsilane, vinyltris 2-methoxyethoxy silane, vinyl tris 2-methoxyethoxy silane, silicone a-172, 6-2-methoxyethoxy-6-vinyl-2,5,7,10-tetraoxa-6-silaundecane, tris methoxyethxy vinylsilane, tris methoxyethoxy vinylsilane, vinyltris methoxyethoxy silane, nuca 172, 2,5,7,10-tetraoxa-6-silaundecane, 6-ethenyl-6-2-methoxyethoxy
PubChem CID 14025
IUPAC-namn ethenyl-tris(2-methoxyethoxy)silane
LEDER COCCO[Si](C=C)(OCCOC)OCCOC
Specifikationer
Kokpunkt 136°C (5.5mmHg)
Kemiskt namn eller material Tris(2-methoxyethoxy)vinylsilane
Fysisk form Liquid
Kvantitet 100 mL
Beilstein 1795316
Flampunkt 113°C (235°F)
Formel vikt 280.39
Procent renhet 98%
Känslighet Moisture sensitive
Densitet 1.034