Läs mer
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex, TRC
Produktkod. 30449504
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30449504 1 mg
1 options
Produktkod. 30449504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8845231MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(hexafluorophosphate) complex
CAS 123148-15-2
Molekylformel C72H48N6Ru • 2[F6P]
Kvantitet 1 mg
Synonym (OC-6-11)-tris(4,7-Diphenyl-1,10-phenanthroline-κN1,κN10)-bis[hexafluorophosphate(1-)] Ruthenium(2+), 1,10-Phenanthroline','4,7-diphenyl-','ruthenium complex (ZCI), Ruthenium(2+)','tris(4,7-diphenyl-1,10-phenanthroline-N1,N10)-','(OC-6-11)-','bis[hexafluorophosphate(1-)] (ZCI), Ruthenium(2+)','tris(4,7-diphenyl-1,10-phenanthroline-κN1,κN10)-','(OC-6-11)-','bis[hexafluorophosphate(1-)] (9CI), Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium bis(hexafluorophosphate)
InChI formel InChI=1S/3C24H16N2.2F6P.Ru/c3*1-3-7-17(8-4-1)19-13-15-25-23-21(19)11-12-22-20(14-16-26-24(22)23)18-9-5-2-6-10-18;2*1-7(2,3,4,5)6;/h3*1-16H;;;/q;;;2*-1;+2
LEDER [F-][P+5]([F-])([F-])([F-])([F-])[F-].C1(C2=C(C=CC3=C4[N]([Ru+2]567([N]8=C9C%10=C(C(C%11=CC=CC=C%11)=CC=[N]7%10)C=CC9=C(C%12=CC=CC=C%12)C=C8)[N]%13=C%14C%15=C(C(C%16=CC=CC=C%16)=CC=[N]6%15)C=CC%14=C(C%17=CC=CC=C%17)C=C%13)=CC=C3C%18=CC=CC=C%18)C4=[N]5C=C2)=CC=CC=C1
Molekylvikt (g/mol) 1388.18
Formel vikt 1388.22