Läs mer
Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(perchlorate), TRC
Produktkod. 30452140
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30452140 500 mg
1 options
Produktkod. 30452140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT884520500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) bis(perchlorate)
CAS 75213-31-9
Molekylformel C72H48N6Ru .2(ClO4)
Kvantitet 500 mg
Synonym Ruthenium(II) tris(4,7-diphenyl-1,10-phenanthroline) Diperchlorate, Ruthenium(II) tris-4,7-diphenyl-1,10-phenanthroline Perchlorate, Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(2+) Diperchlorate, Tris(4,7-diphenyl-1,10-phenanthroline)ruthenium(II) Diperchlorate
InChI formel InChI=1S/C11H9Cl2N3OS/c1-5-6(12)2-3-7(13)9(5)16-10(17)8-4-15-11(14)18-8/h2-4H,1H3,(H2,14,15)(H,16,17)
LEDER Cc1c(Cl)ccc(Cl)c1NC(=O)c2cnc(N)s2
IUPAC-namn 2-amino-N-(3,6-dichloro-2-methylphenyl)-1,3-thiazole-5-carboxamide
Molekylvikt (g/mol) 1297.2
Formel vikt 300.9843