missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tris(hydroxymethyl)aminomethane, Molecular Biology Grade, 99.9%

Catalog Number p-7042016
Click to view available options
Quantity:
100 g
500 g
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
TRIS
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
TRIS
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 168°C to 172°C
Boiling Point 219°C to 220°C (10mmHg)
Color White
Density 1.353
Physical Form Crystalline Powder
Quantity 100 g
Beilstein 741883
Merck Index 14,9772
Solubility Information Freely soluble in water
Formula Weight 121.14
Percent Purity 99.9%
Grade Molecular Biology
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'