missing translation for 'onlineSavingsMsg'
Läs mer

Tris(Hydroxymethyl) Methylamine, Biological Buffer, Fisher Chemical™

Product Code. 10345910 Shop All Fisher Chemical Products
Change view
Click to view available options
Kvantitet:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
10345910 100 g 100g
10274300 500 g 500g
10355910 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10345910 Supplier Fisher Chemical Supplier No. T/P630/48
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 77-86-1
Molekylformel C4H11NO3
Molekylvikt (g/mol) 121.136
MDL-nummer 4679
InChI-nyckel LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC-namn 2-amino-2-(hydroxymethyl)propane-1,3-diol
LEDER C(C(CO)(CO)N)O

Specifications

Smältpunkt 168.5°C
Kokpunkt 219°C
Färg White
pH 10.4
Fysisk form Powder or Solid
Kvantitet 100 g
Förpackning Plastic powder jar
Formel vikt 121.14g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.