Läs mer
Tris(Hydroxymethyl) Methylamine, Biological Buffer, Fisher Chemical™
Produktkod. 10355910 Handla allt Fisher Chemical Produkter
Change view
Kvantitet:
100 g
500 g
1 kg
Förpackningsstorlek:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
10355910 1 kg 1kg
10345910 100 g 100g
10274300 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 10355910 Leverantör Fisher Chemical Leverantörsnummer T/P630/60
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Kemiska identifierare
CAS 77-86-1
Molekylformel C4H11NO3
Molekylvikt (g/mol) 121.136
MDL-nummer 4679
InChI-nyckel LENZDBCJOHFCAS-UHFFFAOYSA-N
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC-namn 2-amino-2-(hydroxymethyl)propane-1,3-diol
LEDER C(C(CO)(CO)N)O
Specifikationer
Smältpunkt 168.5°C
Kokpunkt 219°C
Färg White
pH 10.4
Fysisk form Powder or Solid
Kvantitet 1 kg
Förpackning Plastic powder jar
Formel vikt 121.14g/mol