Läs mer
Tris(tridecyl) Trimellitate, TRC
Produktkod. 30453797
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30453797 1 g
1 options
Produktkod. 30453797 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8864551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Tris(tridecyl) Trimellitate
CAS 94109-09-8
Molekylformel C48 H84 O6
Kvantitet 1 g
Synonym 1,2,4-Benzenetricarboxylic Acid 1,2,4-Tritridecyl Ester, 1,2,4-Benzenetricarboxylic Acid Tritridecyl Ester, Trimellitic Acid Tritridecyl Ester
InChI formel InChI=1S/C48H84O6/c1-4-7-10-13-16-19-22-25-28-31-34-39-52-46(49)43-37-38-44(47(50)53-40-35-32-29-26-23-20-17-14-11-8-5-2)45(42-43)48(51)54-41-36-33-30-27-24-21-18-15-12-9-6-3/h37-38,42H,4-36,39-41H2,1-3H3
LEDER CCCCCCCCCCCCCOC(=O)c1ccc(C(=O)OCCCCCCCCCCCCC)c(c1)C(=O)OCCCCCCCCCCCCC
IUPAC-namn tritridecyl benzene-1,2,4-tricarboxylate
Molekylvikt (g/mol) 757.177
Formel vikt 756.627