Learn More
Tris(trimethylsilyl) phosphite, 92%, Thermo Scientific Chemicals
Catalog Number p-4089618
579.09 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1795-31-9
C9H27O3PSi3
298.54
MFCD00015588
VMZOBROUFBEGAR-UHFFFAOYSA-N
tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
137213
tris(trimethylsilyl) phosphite
C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1795-31-9
C9H27O3PSi3
298.54
MFCD00015588
VMZOBROUFBEGAR-UHFFFAOYSA-N
tris trimethylsilyl phosphite, phosphorous acid tris trimethylsilyl ester, silanol, trimethyl-, phosphite 3:1, tritrimethylsilyl phosphite, tmspi, acmc-209efr, trimethylsilanol phosphite, tris-trimethylsilylphosphite, tris trimethylsily phosphite
137213
tris(trimethylsilyl) phosphite
C[Si](C)(C)OP(O[Si](C)(C)C)O[Si](C)(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8930g/mL
Boiling Point 78.0°C to 81.0°C (8.3 mmHg)
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Linear Formula P[OSi(CH3)3]3
Refractive Index 1.4080 to 1.4110 (20°C, 589nm)
Quantity 5 g
Specific Gravity 0.893
Solubility Information Solubility in water: reacts.
Formula Weight 298.54
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Tris(trimethylsilyl) phosphite
Show More Show Less
compliance-icons
Product Identifier
  • Tris(trimethylsilyl) phosphite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.