missing translation for 'onlineSavingsMsg'
Learn More

Tris, Ultra Pure grade, 99.95% min., MP Biomedicals™

Catalog Number 11413551
Click to view available options
Quantity:
1 kg
5 kg
10 kg
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
Tris base, 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
This item is not returnable. View return policy
77-86-1
C4H11NO3
121.136
MFCD00004679
LENZDBCJOHFCAS-UHFFFAOYSA-N
Tris base, 2-Amino-2-(hydroxymethyl)-1, 3-propanediol
6503
CHEBI:9754
2-amino-2-(hydroxymethyl)propane-1,3-diol
C(C(CO)(CO)N)O
Color White
Physical Form Crystalline Powder
Quantity 10 kg
Assay Percent Range >99.9%
Formula Weight 121.1
Grade Ultrapure
Chemical Name or Material Tris
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'