missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRK fused gene Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11571-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a tetratricopeptide repeat-containing component of the anaphase promoting complex/cyclosome (APC/C), a large E3 ubiquitin ligase that controls cell cycle progression by targeting a number of cell cycle regulators such as B-type cyclins for 26S proteasome-mediated degradation through ubiquitination. The encoded protein is required for the proper ubiquitination function of APC/C and for the interaction of APC/C with transcription coactivators. It also interacts with polyA binding protein and represses internal ribosome entry site-mediated translation. Multiple transcript variants encoding different isoforms have been found for this gene. These differences cause translation initiation at a downstream AUG and result in a shorter protein (isoform b), compared to isoform a.Specifications
TRK fused gene | |
Polyclonal | |
Unconjugated | |
TFG | |
Protein TFG, TF6, TFG, TRK fused gene, TRK fused gene protein, TRKT3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10342, 21787, 360709 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92734 | |
Tfg | |
TRK fused gene Fusion Protein Ag2151 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |